Assay Details
| Gene Symbol: Chga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007693.1 | NP_031719.1 | 6 - 7 | 991 | 58 | |
| GenBank mRNA | AK144504.1 | - | 6 - 7 | 985 | 58 | |
| BC026554.1 | - | 6 - 7 | 975 | 4990941 | 58 | |
| M64278.1 | - | 6 - 7 | 991 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Chga
Genomic Map