Assay Details
| Gene Symbol: Ddx24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020494.3 | NP_065240.2 | 1 - 2 | 87 | 58 | |
| GenBank mRNA | AF214732.1 | - | 1 - 2 | 67 | 58 | |
| AK011012.1 | - | 1 - 2 | 31 | 58 | ||
| AK036538.1 | - | 1 - 2 | 51 | 58 | ||
| AK134339.1 | - | 1 - 2 | 65 | 58 | ||
| AK149810.1 | - | 1 - 2 | 38 | 58 | ||
| AK150778.1 | - | 1 - 2 | 39 | 58 | ||
| AK152551.1 | - | 1 - 2 | 39 | 58 | ||
| AK152928.1 | - | 1 - 2 | 39 | 58 | ||
| AK166360.1 | - | 1 - 2 | 45 | 58 | ||
| AK172557.1 | - | 1 - 2 | 38 | 58 | ||
| BC043721.1 | - | 1 - 2 | 55 | 58 | ||
| CJ141277.1 | - | 1 - 2 | 87 | 58 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF214732.1, AK011012.1, AK134339.1, AK150778.1, AK152551.1, AK152928.1, AK166360.1, NM_020494.3 transcripts.
Set Membership: |
|

Ddx24
Genomic Map