Assay Details
| Gene Symbol: Ddx24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159502.1 | NP_001152974.1 | 7 - 8 | 2442 | 87 | |
| NM_020494.3 | NP_065240.2 | 7 - 8 | 2276 | 87 | ||
| GenBank mRNA | AF214732.1 | - | 7 - 8 | 2256 | 87 | |
| AK149810.1 | - | 7 - 8 | 2229 | 87 | ||
| AK150778.1 | - | 7 - 8 | 2228 | 87 | ||
| AK152551.1 | - | 7 - 8 | 2228 | 87 | ||
| AK152928.1 | - | 7 - 8 | 2228 | 87 | ||
| BC043721.1 | - | 7 - 8 | 2244 | 87 | ||
| BC055048.1 | - | 7 - 8 | 2206 | 6484820 | 87 | |
| BC055317.1 | - | 7 - 8 | 2442 | 5696084 | 87 | |
| U46690.1 | - | 6 | 1050 | 87 | ||
More Information
Set Membership: |
|

Ddx24
Genomic Map