| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT MG-430 PM Array Plate |
1441720_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1441720_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1441720_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0200024786.mm.1 |
NR_038162.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200024786.mm.1 |
NR_038162.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200024786.mm.1 |
TC0200002995.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200024781.mm.1 |
NR_038162.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200024781.mm.1 |
NR_038162.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200024781.mm.1 |
TC0200002995.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200024787.mm.1 |
NR_038162.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200024787.mm.1 |
NR_038162.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200024787.mm.1 |
TC0200002995.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200024788.mm.1 |
NR_038162.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200024788.mm.1 |
NR_038162.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200024788.mm.1 |
TC0200002995.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200024785.mm.1 |
NR_038162.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200024785.mm.1 |
NR_038162.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200024785.mm.1 |
TC0200002995.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1441720_PM_at |
- |
- |
| Mouse Expression Set 430 |
1441720_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1441720_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1441720_at |
- |
- |