Assay Details
| Gene Symbol: Stxbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113569.1 | NP_001107041.1 | 7 - 8 | 720 | 67 | |
| NM_009295.2 | NP_033321.2 | 7 - 8 | 715 | 67 | ||
| GenBank mRNA | AF326545.1 | - | 7 - 8 | 574 | 67 | |
| AF326563.1 | - | 7 - 8 | 574 | 67 | ||
| AK004780.1 | - | 2 - 3 | 439 | 67 | ||
| BC031728.1 | - | 7 - 8 | 647 | 4480978 | 67 | |
| D45903.1 | - | 7 - 8 | 609 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stxbp1
Genomic Map