Assay Details
| Gene Symbol: Lrrc57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159609.1 | NP_001153081.1 | 5 - 6 | 883 | 96 | |
| NM_001159610.1 | NP_001153082.1 | 5 - 6 | 934 | 96 | ||
| NM_001159612.1 | NP_001153084.1 | 5 - 6 | 950 | 96 | ||
| NM_025657.3 | NP_079933.2 | 5 - 6 | 833 | 96 | ||
| NR_027510.1 | 5 - 6 | 941 | 96 | |||
| GenBank mRNA | AK003592.1 | - | 5 - 6 | 851 | 96 | |
| AK015684.1 | - | 5 - 6 | 885 | 96 | ||
| AK134971.1 | - | 5 - 6 | 792 | 96 | ||
| AK169249.1 | - | 5 - 6 | 857 | 96 | ||
| BC034894.1 | - | 5 - 6 | 792 | 3328402 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21576250 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21576252 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21576249 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2037408 | AK169249.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2037408 | BC034894.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2037408 | AK134971.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2037408 | AK012637.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2037408 | NR_027510.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 16703 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 574803 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 651880 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 390069 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 104063 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136241 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2037408 | AK169249.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2037408 | BC034894.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2037408 | AK134971.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2037408 | AK012637.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2037408 | NR_027510.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21576219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21576216 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21576235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21576214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21576218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21576233 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21576237 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21576241 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21576244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21576227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21576239 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21576222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21576209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21576247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21576246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21576249 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21576224 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21576229 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21576211 | - | - |
| Whole Mouse Genome G4122A | A_52_P319073 | AK169249.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P319073 | BC034894.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P319073 | AK033084.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P319073 | AK134971.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P319073 | AK012637.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122226 | AK169249.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122226 | BC034894.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122226 | AK134971.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122226 | AK012637.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122226 | NR_027510.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108825 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01830362 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01830351 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01830353 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01830352 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21576252 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21576249 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21576250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21576247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21576214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21576203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21576249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21576211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21576233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21576208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21576246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21576237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21576244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21576219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21576198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21576222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21576206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21576229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21576242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21576227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21576210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21576235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21576230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21576224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21576239 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576217 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576227 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02192496 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576233 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576218 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576230 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576244 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576216 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576246 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576208 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576214 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576206 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576241 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576247 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576202 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576203 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576209 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576196 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576239 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576249 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576201 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576237 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576221 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576224 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576211 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576210 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576200 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576219 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576229 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10486554 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486557 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486556 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486561 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486559 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486560 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486555 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486558 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486553 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428312_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430057_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10486552 | - | - |
| Mouse Genome 430 2.0 Array | 1428312_at | - | - |
| Mouse Genome 430 2.0 Array | 1430057_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1428312_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430057_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66606_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66606_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200037561.mm.1 | NM_001159609.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037561.mm.1 | NM_001159609.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037561.mm.1 | NM_001159610.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037561.mm.1 | NM_001159610.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037561.mm.1 | NM_001159612.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037561.mm.1 | NM_001159612.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037561.mm.1 | NM_025657.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037561.mm.1 | NM_025657.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037561.mm.1 | NR_027510.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037561.mm.1 | NR_027510.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037561.mm.1 | TC0200004465.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200037562.mm.1 | NM_001159609.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037562.mm.1 | NM_001159609.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037562.mm.1 | NM_001159610.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037562.mm.1 | NM_001159610.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037562.mm.1 | NM_001159612.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037562.mm.1 | NM_001159612.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037562.mm.1 | NM_025657.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037562.mm.1 | NM_025657.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037562.mm.1 | NR_027510.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037562.mm.1 | NR_027510.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037562.mm.1 | TC0200004465.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200037560.mm.1 | NM_001159609.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037560.mm.1 | NM_001159609.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037560.mm.1 | NM_001159610.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037560.mm.1 | NM_001159610.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037560.mm.1 | NM_001159612.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037560.mm.1 | NM_001159612.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037560.mm.1 | NM_025657.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037560.mm.1 | NM_025657.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037560.mm.1 | NR_027510.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037560.mm.1 | NR_027510.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037560.mm.1 | TC0200004465.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428312_PM_at | - | - |
| Mouse Expression Set 430 | 1428312_at | - | - |
| Mouse Expression Set 430 | 1430057_s_at | - | - |
| Mouse Expression Set 430 | 1430057_PM_s_at | - | - |
| Murine Genome U74v2 Set | 160752_at | - | - |
| Murine Genome U74v2 Set | 160752_PM_at | - | - |
| Murine Genome U74v2 Set | 169579_at | - | - |
| Murine Genome U74v2 Set | 169579_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428312_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428312_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428312_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428312_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430057_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430057_PM_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 101090519 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 50037 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000050037 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1251092 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1251092 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1237930 | - | - |
| mouse-6 v1.1 (Illumina) | scl18778.5_150-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 50037 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101090519 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl18778.5_150-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 50037 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101090519 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2898944 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2898944 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1237930 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl18778.5_150-S | - | - |

Lrrc57
Genomic Map