Assay Details
| Gene Symbol: Fcna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007995.3 | NP_032021.1 | 3 - 4 | 400 | 90 | |
| GenBank mRNA | AB007813.1 | - | 3 - 4 | 292 | 90 | |
| AK133539.1 | - | 3 - 4 | 377 | 90 | ||
| AK138752.1 | - | 3 - 4 | 347 | 90 | ||
| AK159651.1 | - | 3 - 4 | 370 | 90 | ||
| BC019180.1 | - | 3 - 4 | 348 | 5008934 | 90 | |
| CT010383.1 | - | 3 - 4 | 262 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fcna
Genomic Map