Assay Details
| Gene Symbol: Pmpca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173180.3 | NP_775272.1 | 11 - 12 | 1282 | 114 | |
| GenBank mRNA | AK004549.1 | - | 11 - 12 | 1282 | 114 | |
| AK075965.1 | - | 11 - 12 | 1282 | 114 | ||
| AK147110.1 | - | 11 - 12 | 1285 | 114 | ||
| AK150118.1 | - | 11 - 12 | 1283 | 114 | ||
| AK158990.1 | - | 11 - 12 | 1250 | 114 | ||
| AK161291.1 | - | 11 - 12 | 1283 | 114 | ||
| AK167975.1 | - | 11 - 12 | 1281 | 114 | ||
| AK169325.1 | - | 11 - 12 | 1277 | 114 | ||
| AK169342.1 | - | 11 - 12 | 1277 | 114 | ||
| BC010810.1 | - | 11 - 12 | 1267 | 4241697 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pmpca
Genomic Map