Assay Details
| Gene Symbol: Gm12292 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK036759.1 | - | 1 - 2 | 72 | 105 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | heparan sulfate (glucosamine) 3-O-sulfotransferase 3A1 | Hs3st3a1 |
3Ost3a, Hs3st3a |
-317.8kb | Anti-sense | Chr. 11 - 64435332 - 64522835 on NCBI Build GRCm38 |
| Downstream | elaC homolog 2 (E. coli) | Elac2 |
1110017O07Rik, D11Wsu80e, Hpc2 |
135.3kb | Anti-sense | Chr. 11 - 64979023 - 65002076 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm12292
Genomic Map