Assay Details
| Gene Symbol: Cd82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136055.2 | NP_001129527.1 | 3 - 4 | 207 | 83 | |
| NM_001271430.1 | NP_001258359.1 | 2 - 3 | 263 | 83 | ||
| NM_001271431.1 | NP_001258360.1 | 2 - 3 | 131 | 83 | ||
| NM_001271432.1 | NP_001258361.1 | 4 - 5 | 426 | 83 | ||
| NM_001271461.1 | NP_001258390.1 | 3 - 4 | 218 | 83 | ||
| NM_001271462.1 | NP_001258391.1 | 3 - 4 | 230 | 83 | ||
| NM_007656.5 | NP_031682.1 | 3 - 4 | 339 | 83 | ||
| GenBank mRNA | AK135381.1 | - | 3 - 4 | 203 | 83 | |
| AK146912.1 | - | 3 - 4 | 203 | 83 | ||
| AK152871.1 | - | 3 - 4 | 334 | 83 | ||
| AK153070.1 | - | 3 - 4 | 334 | 83 | ||
| AK158033.1 | - | 3 - 4 | 282 | 83 | ||
| AK160907.1 | - | 2 - 3 | 185 | 83 | ||
| AK168097.1 | - | 3 - 4 | 290 | 83 | ||
| AK168515.1 | - | 3 - 4 | 309 | 83 | ||
| BC132559.1 | - | 3 - 4 | 309 | 40130836 | 83 | |
| BC132565.1 | - | 3 - 4 | 309 | 40130842 | 83 | |
| D14883.1 | - | 2 - 3 | 172 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd82
Genomic Map