Assay Details
| Gene Symbol: Cd82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136055.2 | NP_001129527.1 | 6 - 7 | 473 | 100 | |
| NM_001271430.1 | NP_001258359.1 | 5 - 6 | 529 | 100 | ||
| NM_001271431.1 | NP_001258360.1 | 5 - 6 | 397 | 100 | ||
| NM_001271432.1 | NP_001258361.1 | 7 - 8 | 692 | 100 | ||
| NM_001271461.1 | NP_001258390.1 | 6 - 7 | 484 | 100 | ||
| NM_001271462.1 | NP_001258391.1 | 6 - 7 | 496 | 100 | ||
| NM_007656.5 | NP_031682.1 | 6 - 7 | 605 | 100 | ||
| GenBank mRNA | AK135381.1 | - | 6 - 7 | 469 | 100 | |
| AK146912.1 | - | 6 - 7 | 469 | 100 | ||
| AK152871.1 | - | 6 - 7 | 600 | 100 | ||
| AK153070.1 | - | 6 - 7 | 600 | 100 | ||
| AK160907.1 | - | 5 - 6 | 451 | 100 | ||
| AK168097.1 | - | 6 - 7 | 556 | 100 | ||
| AK168515.1 | - | 6 - 7 | 575 | 100 | ||
| BC132559.1 | - | 6 - 7 | 575 | 40130836 | 100 | |
| BC132565.1 | - | 6 - 7 | 575 | 40130842 | 100 | |
| D14883.1 | - | 5 - 6 | 438 | 100 | ||
More Information
Set Membership: |
|

Cd82
Genomic Map