Assay Details
| Gene Symbol: Gtf2h5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181392.3 | NP_852057.2 | 1 - 2 | 90 | 74 | |
| GenBank mRNA | AK003547.1 | - | 1 - 2 | 79 | 74 | |
| AK003579.1 | - | 1 - 2 | 74 | 74 | ||
| AK011281.1 | - | 1 - 2 | 75 | 74 | ||
| AK013225.1 | - | 1 - 2 | 54 | 74 | ||
| AK019011.1 | - | 1 - 2 | 45 | 74 | ||
| AK084443.1 | - | 1 - 2 | 82 | 74 | ||
| BC029238.1 | - | 1 - 2 | 66 | 4489531 | 74 | |
| W50582.1 | - | 1 - 2 | 89 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK003547.1, AK003579.1, AK011281.1, AK013225.1, BC029238.1, NM_181392.3 transcripts.
Set Membership: |
|

Gtf2h5
Genomic Map