Assay Details
| Gene Symbol: Cox10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178379.3 | NP_848466.1 | 3 - 4 | 585 | 78 | |
| GenBank mRNA | AK010385.1 | - | 3 - 4 | 579 | 78 | |
| AK149999.1 | - | 3 - 4 | 575 | 78 | ||
| AK154668.1 | - | 3 - 4 | 584 | 78 | ||
| AK171661.1 | - | 3 - 4 | 575 | 78 | ||
| BC018301.1 | - | 3 - 4 | 536 | 78 | ||
| BC038046.1 | - | 3 - 4 | 538 | 4206826 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cox10
Genomic Map