Assay Details
| Gene Symbol: Tcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009337.3 | NP_033363.1 | 2 - 3 | 381 | 73 | |
| GenBank mRNA | AF031956.1 | - | 2 - 3 | 357 | 73 | |
| AK163387.1 | - | 2 - 3 | 364 | 73 | ||
| BC052336.1 | - | 2 - 3 | 362 | 30053730 | 73 | |
| DV653827.1 | - | 2 - 3 | 381 | 73 | ||
| DV663230.1 | - | 1 - 2 | 83 | 73 | ||
| Y15376.1 | - | 2 - 3 | 357 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tcl1
Genomic Map