Assay Details
| Gene Symbol: Moap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142937.2 | NP_001136409.1 | 1 - 2 | 120 | 64 | |
| GenBank mRNA | AU080670.1 | - | 1 - 2 | 104 | 64 | |
| BY741099.1 | - | 1 - 2 | 118 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001142937.2 transcripts.
Set Membership: |
|

Moap1
Genomic Map