Assay Details
| Gene Symbol: Nol7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023554.2 | NP_076043.2 | 4 - 5 | 428 | 77 | |
| GenBank mRNA | AB041566.1 | - | 4 - 5 | 412 | 77 | |
| AK008670.1 | - | 4 - 5 | 413 | 77 | ||
| AK019972.1 | - | 4 - 5 | 413 | 77 | ||
| AK032963.1 | - | 4 - 5 | 414 | 77 | ||
| BC090637.1 | - | 4 - 5 | 401 | 30940523 | 77 | |
| BC130012.1 | - | 4 - 5 | 418 | 40140312 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nol7
Genomic Map