Assay Details
| Gene Symbol: Nol7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023554.2 | NP_076043.2 | 5 - 6 | 510 | 121 | |
| GenBank mRNA | AB041566.1 | - | 5 - 6 | 494 | 121 | |
| AK008670.1 | - | 5 - 6 | 495 | 121 | ||
| AK019972.1 | - | 5 - 6 | 495 | 121 | ||
| AK032963.1 | - | 5 - 6 | 496 | 121 | ||
| BC090637.1 | - | 5 - 6 | 483 | 30940523 | 121 | |
| BC130012.1 | - | 5 - 6 | 500 | 40140312 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28989931 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28989933 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28989935 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28989932 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28989936 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28989934 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P152612 | AK032963.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P152612 | AK140839.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P152612 | BC090637.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P152612 | NM_023554.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P152612 | AB041566.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P152612 | AK032963.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P152612 | AK140839.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P152612 | BC090637.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P152612 | NM_023554.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P152612 | AB041566.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923638 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 830286 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 28743 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P126965 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P108397 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P152612 | AK032963.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P152612 | AK140839.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P152612 | BC090637.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P152612 | NM_023554.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P152612 | AB041566.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P152612 | AK032963.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P152612 | AK140839.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P152612 | BC090637.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P152612 | NM_023554.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P152612 | AB041566.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28989918 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28989943 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28989920 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28989941 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28989950 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28989940 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28989916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28989934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28989926 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28989951 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28989936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28989949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28989928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28989935 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28989931 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28989945 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28989938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28989947 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28989923 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28989948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28989930 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28989933 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28989924 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07854870 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P108397 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P126965 | - | - |
| Whole Mouse Genome G4122A | A_51_P152612 | AK032963.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P152612 | AK140839.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P152612 | BC090637.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P152612 | NM_023554.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P152612 | AB041566.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P152612 | AK032963.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P152612 | AK140839.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P152612 | BC090637.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P152612 | NM_023554.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P152612 | AB041566.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P609049 | AK032963.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P609049 | BC090637.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P609049 | BC130012.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P609049 | NM_023554.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P609049 | AB041566.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128505 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P126965 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P108397 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10463715 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10858129 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10858131 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10754856 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10971297 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28989931 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28989936 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28989932 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28989933 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28989935 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28989934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28989964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28989936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28989916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28989950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28989920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28989966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28989945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28989927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28989967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28989928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28989943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28989965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28989947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28989938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28989933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28989960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28989930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28989959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28989925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28989953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28989955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28989926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28989948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28989941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28989931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28989923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28989934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28989946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28989933 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28989955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28989936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28989949 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28989924 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28989927 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28989965 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28989964 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28989938 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10497482 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28989930 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28989918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28989956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28989943 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10971297 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28989966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28989919 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28989935 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10775562 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28989950 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28989945 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10855062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28989959 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28989953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10463715 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28989967 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28989928 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28989920 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10859372 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28989948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28989923 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28989925 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10404829 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404830 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404834 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404828 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404832 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404835 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404831 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404833 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1458059_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420486_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420485_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420487_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10404827 | - | - |
| Mouse Genome 430 2.0 Array | 1420486_at | - | - |
| Mouse Genome 430 2.0 Array | 1420485_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1458059_at | - | - |
| Mouse Genome 430 2.0 Array | 1458059_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448064_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448064_at | - | - |
| Mouse Genome 430 2.0 Array | 1420486_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1420487_at | - | - |
| Mouse Genome 430 2.0 Array | 1420485_at | - | - |
| Mouse Genome 430 2.0 Array | 1420487_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70078_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70078_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300002983.mm.1 | NM_023554.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002983.mm.1 | NM_023554.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002983.mm.1 | TC1300000491.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300002980.mm.1 | NM_023554.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002980.mm.1 | NM_023554.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002980.mm.1 | TC1300000491.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300002982.mm.1 | NM_023554.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002982.mm.1 | NM_023554.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002982.mm.1 | TC1300000491.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300002979.mm.1 | NM_023554.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002979.mm.1 | NM_023554.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002979.mm.1 | TC1300000491.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300002984.mm.1 | NM_023554.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002984.mm.1 | NM_023554.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002984.mm.1 | TC1300000491.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300002985.mm.1 | NM_023554.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002985.mm.1 | NM_023554.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002985.mm.1 | TC1300000491.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1420487_PM_at | - | - |
| Mouse Expression Set 430 | 1420485_at | - | - |
| Mouse Expression Set 430 | 1420487_at | - | - |
| Mouse Expression Set 430 | 1420485_PM_at | - | - |
| Mouse Expression Set 430 | 1420486_PM_at | - | - |
| Mouse Expression Set 430 | 1420486_at | - | - |
| Mouse Expression Set 430 | 1458059_PM_at | - | - |
| Mouse Expression Set 430 | 1458059_at | - | - |
| Murine Genome U74v2 Set | 160398_at | - | - |
| Murine Genome U74v2 Set | 160398_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420485_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420485_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420486_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420487_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420486_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420487_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420487_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420485_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420485_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420486_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420486_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420487_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458059_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458059_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 535367 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6520332 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6520332 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006520332 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1250073 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1250073 | - | - |
| mouse-6 v1.1 (Illumina) | scl44846.7_0-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6520332 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl44846.7_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6520332 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1250073 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1250073 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl44846.7_0-S | - | - |

Nol7
Genomic Map