Assay Details
| Gene Symbol: Creb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK084024.1 | - | 445 | 75 | ||
| AK170766.1 | - | 519 | 75 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Creb5| Gene Symbol: Creb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK084024.1 | - | 445 | 75 | ||
| AK170766.1 | - | 519 | 75 | |||
Set Membership: |
|