Assay Details
| Gene Symbol: Creb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172728.2 | NP_766316.1 | 2 - 3 | 194 | 63 | |
| GenBank mRNA | AK052230.1 | - | 2 - 3 | 197 | 63 | |
| AK052451.1 | - | 2 - 3 | 195 | 63 | ||
| AK084024.1 | - | 575 | 63 | |||
| AK154148.1 | - | 2 - 3 | 195 | 63 | ||
| AK155537.1 | - | 2 - 3 | 192 | 63 | ||
| AK170128.1 | - | 2 - 3 | 195 | 63 | ||
| AK170766.1 | - | 649 | 63 | |||
| BC029022.1 | - | 1 - 2 | 129 | 5012117 | 63 | |
| BC098487.1 | - | 2 - 3 | 184 | 30090687 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Creb5
Genomic Map