Assay Details
| Gene Symbol: Creb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK154148.1 | - | 4 - 5 | 625 | 60 | |
| AK155537.1 | - | 4 - 5 | 622 | 60 | ||
| BC029022.1 | - | 3 - 4 | 559 | 5012117 | 60 | |
| BC098487.1 | - | 4 - 5 | 614 | 30090687 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Creb5
Genomic Map