Assay Details
| Gene Symbol: Ccdc126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC095987.1 | - | 1 - 2 | 370 | 5365989 | 93 |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc126| Gene Symbol: Ccdc126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC095987.1 | - | 1 - 2 | 370 | 5365989 | 93 |
Set Membership: |
|