Assay Details
| Gene Symbol: Hoxa11os | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | U20367.1 | - | LIT1564 Hoxa11as |
1 - 2 | 575 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | homeobox A11 | Hoxa11 |
Hox-1.9, Hoxa-11 |
Anti-sense | Chr. 6 - 52242105 - 52245810 on NCBI Build GRCm38 | |
| Downstream | homeobox A13 | Hoxa13 |
Hd, Hox-1.10 |
12.0kb | Anti-sense | Chr. 6 - 52258853 - 52260880 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Hoxa11os
Genomic Map