Assay Details
| Gene Symbol: Nudt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016918.3 | NP_058614.1 | 5 - 6 | 680 | 64 | |
| GenBank mRNA | AF222786.1 | - | 4 - 5 | 330 | 64 | |
| AK088222.1 | - | 5 - 6 | 681 | 64 | ||
| BC049595.1 | - | 5 - 6 | 485 | 6774692 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nudt5
Genomic Map