Assay Details
| Gene Symbol: Il15ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271497.1 | NP_001258426.1 | 5 - 6 | 693 | 65 | |
| NM_001271499.1 | NP_001258428.1 | 5 - 6 | 657 | 65 | ||
| NM_001271500.1 | NP_001258429.1 | 4 - 5 | 558 | 65 | ||
| NM_001271501.1 | NP_001258430.1 | 3 - 4 | 465 | 65 | ||
| NM_008358.2 | NP_032384.1 | 5 - 6 | 756 | 65 | ||
| NM_133836.2 | NP_598597.1 | 5 - 6 | 809 | 65 | ||
| NR_073191.1 | 4 - 5 | 611 | 65 | |||
| NR_073192.1 | 3 - 4 | 518 | 65 | |||
| GenBank mRNA | AY219715.1 | - | 5 - 6 | 558 | 65 | |
| AY219716.1 | - | 4 - 5 | 460 | 65 | ||
| AY219717.1 | - | 3 - 4 | 367 | 65 | ||
| AY221616.1 | - | 5 - 6 | 434 | 65 | ||
| AY221617.1 | - | 4 - 5 | 352 | 65 | ||
| AY221618.1 | - | 3 - 4 | 259 | 65 | ||
| BC022705.1 | - | 5 - 6 | 566 | 65 | ||
| BC069881.1 | - | 4 - 5 | 517 | 65 | ||
| BC095982.1 | - | 2 - 3 | 484 | 65 | ||
| BC132231.1 | - | 4 - 5 | 356 | 65 | ||
| BC132233.1 | - | 4 - 5 | 356 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AY221617.1, AY221618.1 transcripts.
Set Membership: |
|

Il15ra
Genomic Map