Assay Details
| Gene Symbol: Stam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011484.2 | NP_035614.1 | 9 - 10 | 1091 | 149 | |
| GenBank mRNA | AK035924.1 | - | 9 - 10 | 1014 | 149 | |
| AK144987.1 | - | 9 - 10 | 1108 | 149 | ||
| AK147832.1 | - | 9 - 10 | 1001 | 149 | ||
| AK157767.1 | - | 9 - 10 | 1038 | 149 | ||
| AK162431.1 | - | 9 - 10 | 1038 | 149 | ||
| AK162973.1 | - | 4 - 5 | 276 | 149 | ||
| AK163720.1 | - | 9 - 10 | 1041 | 149 | ||
| AK163922.1 | - | 9 - 10 | 1028 | 149 | ||
| AK169170.1 | - | 9 - 10 | 1098 | 149 | ||
| BC044666.1 | - | 9 - 10 | 1091 | 4950569 | 149 | |
| BC055326.1 | - | 9 - 10 | 1037 | 6416552 | 149 | |
| U43900.1 | - | 9 - 10 | 948 | 149 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stam
Genomic Map