Assay Details
| Gene Symbol: Golga2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080968.1 | NP_001074437.1 | 18 - 19 | 1852 | 65 | |
| NM_133852.2 | NP_598613.2 | 19 - 20 | 1933 | 65 | ||
| GenBank mRNA | AK076370.1 | - | 13 - 14 | 1551 | 65 | |
| AK077516.1 | - | 18 - 19 | 1857 | 65 | ||
| AK220509.1 | - | 19 - 20 | 1861 | 65 | ||
| BC011407.1 | - | 17 - 18 | 1653 | 3596079 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Golga2
Genomic Map