Assay Details
| Gene Symbol: Golga2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080968.1 | NP_001074437.1 | 21 - 22 | 2258 | 65 | |
| NM_133852.2 | NP_598613.2 | 22 - 23 | 2339 | 65 | ||
| GenBank mRNA | AK076370.1 | - | 16 - 17 | 1957 | 65 | |
| AK220509.1 | - | 22 - 23 | 2267 | 65 | ||
| BC011407.1 | - | 20 - 21 | 2059 | 3596079 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Golga2
Genomic Map