Assay Details
| Gene Symbol: Golga2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133852.2 | NP_598613.2 | 3 - 4 | 352 | 82 | |
| GenBank mRNA | AK220509.1 | - | 3 - 4 | 280 | 82 | |
More Information
Set Membership: |
|

Golga2
Genomic Map