Assay Details
| Gene Symbol: Golga2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080968.1 | NP_001074437.1 | 3 - 4 | 363 | 81 | |
| NM_133852.2 | NP_598613.2 | 4 - 5 | 444 | 81 | ||
| GenBank mRNA | AK220509.1 | - | 4 - 5 | 372 | 81 | |
| BB855623.1 | - | 3 - 4 | 270 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Golga2
Genomic Map