Assay Details
| Gene Symbol: Golga2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080968.1 | NP_001074437.1 | 6 - 7 | 530 | 69 | |
| NM_133852.2 | NP_598613.2 | 7 - 8 | 611 | 69 | ||
| GenBank mRNA | AK076370.1 | - | 2 - 3 | 151 | 69 | |
| AK077516.1 | - | 6 - 7 | 531 | 69 | ||
| AK220509.1 | - | 7 - 8 | 539 | 69 | ||
| BB855623.1 | - | 6 - 7 | 437 | 69 | ||
| BC011407.1 | - | 5 - 6 | 331 | 3596079 | 69 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC011407.1 transcripts.
Set Membership: |
|

Golga2
Genomic Map