Assay Details
| Gene Symbol: Golga2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080968.1 | NP_001074437.1 | 2 - 3 | 269 | 82 | |
| GenBank mRNA | AK077516.1 | - | 2 - 3 | 270 | 82 | |
| BB855623.1 | - | 2 - 3 | 176 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Golga2
Genomic Map