Assay Details
| Gene Symbol: Dolpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290508.1 | NP_001277437.1 | 2 - 3 | 464 | 129 | |
| GenBank mRNA | AK002544.1 | - | 2 - 3 | 303 | 129 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002544.1, NM_001290508.1 transcripts.
Set Membership: |
|

Dolpp1
Genomic Map