Assay Details
| Gene Symbol: Ccbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172404.2 | NP_765992.2 | 2 - 3 | 269 | 53 | |
| GenBank mRNA | BC016206.1 | - | 2 - 3 | 371 | 4912335 | 53 |
| BC025124.1 | - | 1 - 2 | 402 | 4486058 | 53 | |
| BC052047.1 | - | 2 - 3 | 269 | 5687475 | 53 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC016206.1, BC025124.1, BC052047.1, NM_172404.2 transcripts.
Set Membership: |
|

Ccbl1
Genomic Map