Assay Details
| Gene Symbol: Ak8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033874.2 | NP_001029046.2 | 11 - 12 | 1153 | 77 | |
| GenBank mRNA | AB046266.1 | - | 2 - 3 | 172 | 77 | |
| BC100481.1 | - | 11 - 12 | 1150 | 30917849 | 77 | |
| BC109352.2 | - | 11 - 12 | 1140 | 40057006 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ak8
Genomic Map