Assay Details
| Gene Symbol: Ak8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033874.2 | NP_001029046.2 | 5 - 6 | 433 | 139 | |
| GenBank mRNA | AK004439.1 | - | 5 - 6 | 431 | 139 | |
| BC100481.1 | - | 5 - 6 | 430 | 30917849 | 139 | |
| BC109352.2 | - | 5 - 6 | 420 | 40057006 | 139 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ak8
Genomic Map