Assay Details
| Gene Symbol: Ak8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033874.2 | NP_001029046.2 | 8 - 9 | 789 | 58 | |
| GenBank mRNA | BC100481.1 | - | 8 - 9 | 786 | 30917849 | 58 |
| BC109352.2 | - | 8 - 9 | 776 | 40057006 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ak8
Genomic Map