Assay Details
| Gene Symbol: Gtf3c5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290484.1 | NP_001277413.1 | 9 - 10 | 1548 | 68 | |
| NM_148928.3 | NP_683730.1 | 9 - 10 | 1548 | 68 | ||
| GenBank mRNA | AK045419.1 | - | 9 - 10 | 1140 | 68 | |
| AK147014.1 | - | 9 - 10 | 1526 | 68 | ||
| BC023847.1 | - | 2 - 3 | 159 | 68 | ||
| BC027055.1 | - | 9 - 10 | 1515 | 5360397 | 68 | |
| BC027247.1 | - | 9 - 10 | 1458 | 2631223 | 68 | |
| BC039171.1 | - | 9 - 10 | 1469 | 4500799 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gtf3c5
Genomic Map