Assay Details
| Gene Symbol: Exoc3l4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289487.1 | NP_001276416.1 | 7 - 8 | 1820 | 55 | |
| NM_001289488.1 | NP_001276417.1 | 7 - 8 | 1571 | 55 | ||
| NM_001289489.1 | NP_001276418.1 | 6 - 7 | 1466 | 55 | ||
| NM_028807.4 | NP_083083.3 | 6 - 7 | 1715 | 55 | ||
| GenBank mRNA | AK004674.1 | - | 7 - 8 | 1572 | 55 | |
| AK005443.1 | - | 5 - 6 | 1462 | 55 | ||
| BC042799.1 | - | 3 - 4 | 455 | 55 | ||
| BC075664.1 | - | 6 - 7 | 1556 | 30604610 | 55 | |
More Information
Updated Mapping Information
Set Membership: |
|

Exoc3l4
Genomic Map