Assay Details
| Gene Symbol: Ccdc73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177600.4 | NP_808268.2 | 14 - 15 | 1380 | 164 | |
| GenBank mRNA | AK029862.1 | - | 6 - 7 | 587 | 164 | |
| BC095976.1 | - | 13 - 14 | 1126 | 164 | ||
| BC150667.1 | - | 5 - 6 | 509 | 9087578 | 164 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc73
Genomic Map