Assay Details
| Gene Symbol: Ccdc73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177600.4 | NP_808268.2 | 3 - 4 | 403 | 140 | |
| GenBank mRNA | BB615231.1 | - | 3 - 4 | 401 | 138 | |
| BC095976.1 | - | 2 - 3 | 149 | 140 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC095976.1 transcripts.
Set Membership: |
|

Ccdc73
Genomic Map