Assay Details
| Gene Symbol: Ccdc73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177600.4 | NP_808268.2 | 4 - 5 | 476 | 74 | |
| GenBank mRNA | BB615231.1 | - | 4 - 5 | 474 | 74 | |
| BC095976.1 | - | 3 - 4 | 222 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC095976.1 transcripts.
Set Membership: |
|

Ccdc73
Genomic Map