Assay Details
| Gene Symbol: Spata31d1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028157.2 | NP_082433.2 | 3 - 4 | 331 | 74 | |
| GenBank mRNA | AK029673.1 | - | 3 - 4 | 316 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK029673.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P270158 | AK029673.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P270158 | NM_028157.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P270158 | CR520895.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P270158 | AK029673.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P270158 | NM_028157.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P270158 | CR520895.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 604893 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596930 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 121556 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P126561 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P270158 | AK029673.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P270158 | NM_028157.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P270158 | CR520895.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P270158 | AK029673.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P270158 | NM_028157.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P270158 | CR520895.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081097 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081084 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081063 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081090 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081080 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081098 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081088 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081077 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081104 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081093 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081106 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081089 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081076 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081075 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081102 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081099 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081069 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081086 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081083 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29081095 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P126561 | - | - |
| Whole Mouse Genome G4122A | A_52_P270158 | AK029673.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P270158 | NM_028157.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P270158 | CR520895.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P270158 | AK029673.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P270158 | NM_028157.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P270158 | CR520895.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102167 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P126561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29080981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29080963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29080999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29080988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29080982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29080962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29080995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29080970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29080978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29080986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29080991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29080979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29080984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29080972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29080998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29080993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29080966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29080975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29080974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29080959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29080985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29081061 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081063 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081104 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081023 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081117 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081131 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081037 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10652666 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081066 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10993250 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081075 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081122 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081089 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10938926 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10993853 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081077 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081119 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081098 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081082 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081059 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081108 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081053 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081047 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081051 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081048 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081102 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081107 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10678097 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081080 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081124 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081127 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10938920 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081097 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081090 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081109 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081106 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10938900 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10448590 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081052 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081130 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081085 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081125 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081057 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081036 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11066549 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081084 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29081039 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10409788 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409789 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409787 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10409786 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300013606.mm.1 | NM_028157.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013606.mm.1 | NM_028157.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013606.mm.1 | TC1300002121.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 312998 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4590735 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004590735 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1216199 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1216199 | - | - |
| mouse-6 v1.1 (Illumina) | scl43877.4_668-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4590735 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl43877.4_668-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4590735 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2630401 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2630401 | - | - |

Spata31d1a
Genomic Map