Assay Details
| Gene Symbol: Gpr158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004761.1 | NP_001004761.1 | 4 - 5 | 2025 | 96 | |
| GenBank mRNA | AB093287.1 | - | 4 - 5 | 1130 | 96 | |
| AK030530.1 | - | 4 - 5 | 2021 | 96 | ||
| AK083234.1 | - | 4 - 5 | 2051 | 96 | ||
| AK086256.1 | - | 4 - 5 | 2026 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpr158
Genomic Map