Assay Details
| Gene Symbol: Arhgap15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153820.3 | NP_722542.2 | 9 - 10 | 903 | 106 | |
| GenBank mRNA | AK036520.1 | - | 9 - 10 | 903 | 106 | |
| AK040796.1 | - | 10 - 11 | 973 | 106 | ||
| AK170537.1 | - | 9 - 10 | 901 | 106 | ||
| BC024887.1 | - | 9 - 10 | 857 | 3156334 | 106 | |
| BC034881.1 | - | 9 - 10 | 876 | 3468639 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap15
Genomic Map