Assay Details
| Gene Symbol: Etl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177631.1 | NP_001171102.1 | 6 - 7 | 2028 | 97 | |
| NM_029895.4 | NP_084171.2 | 7 - 8 | 2241 | 97 | ||
| NM_178059.5 | NP_835160.2 | 7 - 8 | 2241 | 97 | ||
| GenBank mRNA | AB093289.2 | - | 6 - 7 | 1818 | 97 | |
| AB125594.2 | - | 7 - 8 | 2241 | 97 | ||
| AB125595.2 | - | 7 - 8 | 2241 | 97 | ||
| AK054453.1 | - | 2 - 3 | 1184 | 97 | ||
| AK086078.1 | - | 2 - 3 | 1186 | 97 | ||
| AK147510.1 | - | 6 - 7 | 1935 | 97 | ||
| BC172143.1 | - | 6 - 7 | 1748 | 97 | ||
More Information
Set Membership: |
|

Etl4
Genomic Map