Assay Details
| Gene Symbol: Glp2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175681.3 | NP_783612.2 | 8 - 9 | 1153 | 111 | |
| GenBank mRNA | AK035702.1 | - | 548 | 111 | ||
| AY605231.1 | - | 8 - 9 | 872 | 111 | ||
| BC044746.2 | - | 8 - 9 | 1153 | 5363416 | 111 | |
More Information
Set Membership: |
|

Glp2r
Genomic Map