Assay Details
| Gene Symbol: Catsperb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK040480.1 | - | 5 - 6 | 743 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
kinesin family member 4, pseudogene |
Kif4-ps |
4930463M05Rik, Kif4b |
-392.1kb | Sense | Chr. 12 - 101145609 - 101149273 on NCBI Build GRCm38 |
| Downstream |
tandem C2 domains, nuclear |
Tc2n |
4933406D09Rik, Mtac2d1, Tac2-N |
96.0kb | Anti-sense | Chr. 12 - 101645446 - 101719000 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Catsperb
Genomic Map