Assay Details
| Gene Symbol: 4930455J16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK030052.1 | - | 3 - 4 | 412 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 28 (sodium-coupled nucleoside transporter), member 3 |
Slc28a3 | Cnt3 | -145.2kb | Sense | Chr. 13 - 58553307 - 58610877 on NCBI Build GRCm38 |
| Downstream |
neurotrophic tyrosine kinase, receptor, type 2 |
Ntrk2 |
GP145-TrkB/GP95-TrkB, Tkrb, trk-B, trkB |
49.6kb | Anti-sense | Chr. 13 - 58806569 - 59133970 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

4930455J16Rik
Genomic Map