Assay Details
| Gene Symbol: Gm16876 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK040110.1 | - | 504 | 118 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | stonin 2 | Ston2 |
4933401N24Rik, AI425999, AW544599 |
Anti-sense | Chr. 12 - 91633009 - 91786436 on NCBI Build GRCm38 | |
| Downstream | sel-1 suppressor of lin-12-like (C. elegans) | Sel1l |
AW493766, Sel1h, mKIAA4137 |
112.5kb | Anti-sense | Chr. 12 - 91806043 - 91849157 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm16876
Genomic Map