Assay Details
| Gene Symbol: Zim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF401983.1 | - | 240 | 86 | ||
| AK007205.1 | - | 1 - 2 | 220 | 86 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | olfactory receptor 1350 | Olfr1350 | MOR222-3 | -84.0kb | Anti-sense | Chr. 7 - 6569993 - 6570919 on NCBI Build GRCm38 |
| Downstream |
zinc finger, imprinted 1 |
Zim1 | 16.1kb | Sense | Chr. 7 - 6674444 - 6696538 on NCBI Build GRCm38 |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1970349 | AF401983.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 83322 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 843467 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 326405 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 179562 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 43061 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1970349 | AF401983.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01477380 | - | - |
| Whole Mouse Genome G4122A | A_52_P137058 | AF401983.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P137058 | AK007205.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06019741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06062768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24956258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06019734 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06062762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24956243 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24956237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24956260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06176417 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06019730 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24956272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06568717 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24956265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06019738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24956251 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06176415 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24956209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24956277 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24956253 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06005005 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24956238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24956208 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24956215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06639173 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24956234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24956268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24956256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06062761 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06005006 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24956271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24956222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24956274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24956275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24956255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24956201 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06219627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06062767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06305816 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24956241 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24956226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24956235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06019733 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05953023 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24956229 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24956236 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06305815 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06569240 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24956230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24956273 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1432297_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1432297_at | - | - |
| Mouse Genome 430 2.0 Array | 1432297_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76637_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76637_at | - | - |
| Mouse Expression Set 430 | 1432297_PM_at | - | - |
| Mouse Expression Set 430 | 1432297_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432297_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432297_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 106420450 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106420450 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2446073 | - | - |
| mouse-6 v1.1 (Illumina) | scl31775.5.1_265-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106420450 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106420450 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2446073 | - | - |

Zim2
Genomic Map